Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018543.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018543.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018543.hg.1 |
TC0X001392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018548.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018548.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018548.hg.1 |
TC0X001392.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000362227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000362227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010872.hg.1 |
NR_029946.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173794.hg.1 |
TC0X00010872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173806.hg.1 |
TC0X00010872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000362227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000362227.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000414769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000440570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000441492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000445415.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
ENST00000457876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_024607.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010872.hg.1 |
NR_029946.1 |
Refseq |