This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03680373_m1
See other WEE2-AS1 GE Assays ›
Gene Symbol:
WEE2-AS1
Gene Name:
WEE2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 141704338 - 141738230 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03680373_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WEE2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015392.1 5 - 6 647 105
GenBank mRNA AK098171.1 - 5 - 6 647 105
BC101359.2 - 5 - 6 901 105
BC131564.1 - 3 - 4 277 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB026068.1 - 3 - 4 Chr. 7 - 141714235 - 141738346 [-] on Build GRCh38 104
GenBankmRNA DB044037.1 - 3 - 4 288 Chr. 7 - 141705703 - 141738230 [-] on Build GRCh38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream WEE1 homolog 2 (S. pombe) WEE2 WEE1B Anti-sense Chr. 7 - 141708353 - 141731271 on NCBI Build GRCh38
Downstream single stranded DNA binding protein 1 SSBP1 Mt-SSB,
SOSS-B1,
SSBP,
mtSSB
14.3kb Anti-sense Chr. 7 - 141738309 - 141750488 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0