This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03680062_m1
See other LGALS3 GE Assays ›
Gene Symbol:
LGALS3
Gene Name:
lectin, galactoside binding soluble 3
Gene Aliases:
CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2
Chromosome Location:
Chr.14: 55129217 - 55145430 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03680062_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LGALS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177388.1 NP_001170859.1 4 - 5 520 88
NM_002306.3 NP_002297.2 4 - 5 520 88
NR_003225.2 2 - 3 1476 88
XM_011536759.2 XP_011535061.2 5 - 6 510 88
GenBank mRNA AB006780.1 - 4 - 5 485 88
AB209391.1 - 3 - 4 4563 88
AK314929.1 - 4 - 5 519 88
BC001120.2 - 4 - 5 456 3050135 88
BC053667.1 - 4 - 5 471 6148401 88
BC068068.1 - 4 - 5 466 6428428 88
BG332134.1 - 4 - 5 478 88
BQ645629.1 - 583 88
BX161455.1 - 2 - 3 1271 88
BX641090.1 - 2 - 3 1477 88
CD518618.1 - 4 - 5 520 88
CR456897.1 - 3 - 4 432 88
CR542097.1 - 3 - 4 432 88
M35368.1 - 4 - 5 454 88
M57710.1 - 3 - 4 450 88
M64303.1 - 3 - 4 450 88
S59012.1 - 3 - 4 432 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BX161455.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LGALS3

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

monocyte chemotaxis
mRNA processing
RNA splicing
neutrophil chemotaxis
epithelial cell differentiation
regulation of T cell proliferation
innate immune response
negative regulation of endocytosis
eosinophil chemotaxis
macrophage chemotaxis
negative regulation of T cell receptor signaling pathway
positive chemotaxis
regulation of T cell apoptotic process
mononuclear cell migration
positive regulation of mononuclear cell migration
positive regulation of calcium ion import
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of immunological synapse formation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
positive regulation of dendritic cell differentiation
negative regulation of extrinsic apoptotic signaling pathway
protein binding
IgE binding
carbohydrate binding
chemoattractant activity
laminin binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0