This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03677983_m1
See other C1RL-AS1 GE Assays ›
Gene Symbol:
C1RL-AS1
Gene Name:
C1RL antisense RNA 1
Gene Aliases:
MATL2963
Chromosome Location:
Chr.12: 7108308 - 7121851 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03677983_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C1RL-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026947.1 10 - 11 5010 64
GenBank mRNA AK091792.1 - 7 - 8 1562 64
AL833364.1 - 10 - 11 5010 64
AY358730.1 - 2 - 3 73 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream retinol binding protein 5 RBP5 CRBP-III,
CRBP3,
CRBPIII,
HRBPiso
Anti-sense Chr. 12 - 7119106 - 7130272 on NCBI Build GRCh38
Downstream calsyntenin 3 CLSTN3 CDHR14,
CSTN3,
alcbeta
9.7kb Sense Chr. 12 - 7130371 - 7158945 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200006667.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000106.hg.1

Additional Information:

This assay is in the 5'UTR region of the AY358730.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000382215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000435921.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000535078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000535078.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000536679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000541775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000545775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000545775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000545775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 ENST00000545775.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006667.hg.1 NR_026947.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0