Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002664.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002664.hg.1 |
TC6_cox_hap2000154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002601.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002601.hg.1 |
TC6_dbb_hap3000143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002332.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002332.hg.1 |
TC6_mcf_hap5000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002183.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002183.hg.1 |
TC6_mann_hap4000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002331.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002331.hg.1 |
TC6_mcf_hap5000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002186.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4002186.hg.1 |
TC6_mann_hap4000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002640.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002640.hg.1 |
TC6_qbl_hap6000144.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017299.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017299.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017299.hg.1 |
TC06001522.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017300.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017300.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06017300.hg.1 |
TC06001522.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002324.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002324.hg.1 |
TC6_ssto_hap7000126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002600.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002600.hg.1 |
TC6_dbb_hap3000143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002325.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002325.hg.1 |
TC6_ssto_hap7000126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002665.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002665.hg.1 |
TC6_cox_hap2000154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002641.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002641.hg.1 |
TC6_qbl_hap6000144.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0600177032.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177032.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177032.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177032.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177032.hg.1 |
TC0600011407.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600177036.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177036.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177036.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600177036.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600177036.hg.1 |
TC0600011407.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
ENST00000412143.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011407.hg.1 |
NR_026816.1 |
Refseq |