Assay Details
Gene Symbol: MGC16025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026664.1 | 1 | 1648 | 80 | ||
GenBank mRNA | BC008026.1 | - | 1 | 1648 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4441 | MIR4441 | -107.4kb | Sense | Chr. 2 - 239085827 - 239085926 on NCBI Build GRCh38 | |
Downstream | microRNA 4269 | MIR4269 | 110.0kb | Anti-sense | Chr. 2 - 239305462 - 239305545 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P79572 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P79572 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 17748 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36034 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P79572 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P79572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996490 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996497 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996514 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399697 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996514 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996497 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996516 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996490 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399677 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15399692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996513 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01996479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22124552 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P79572 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P79572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996497 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996521 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996510 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399682 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399690 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996494 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399696 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399687 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996502 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399686 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399698 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996491 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996533 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996512 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996482 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996519 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996479 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996525 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996487 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399680 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399684 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996539 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996531 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399703 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996541 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399671 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996516 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399679 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996504 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996527 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15399702 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996500 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996484 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996535 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01996508 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8060062 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8060061 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553747_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553747_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553747_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553747_at2 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015177 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015177 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1757561 | BC008026.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1757561 | NR_026664.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757561 | BC008026.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757561 | NR_026664.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1757561 | BC008026.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1757561 | NR_026664.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757561 | BC008026.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757561 | NR_026664.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757561 | BC008026.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757561 | NR_026664.1 | human_transcriptome |
