This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03676289_s1
See other CHRNA5 GE Assays ›
Gene Symbol:
CHRNA5
Gene Name:
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases:
LNCR2
Chromosome Location:
Chr.15: 78565520 - 78595269 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03676289_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHRNA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC050469.1 - 1 2049 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream proteasome subunit alpha 4 PSMA4 HC9,
HsT17706,
PSC9
-39.9kb Anti-sense Chr. 15 - 78540405 - 78549220 on NCBI Build GRCh38
Downstream cholinergic receptor nicotinic alpha 3 subunit CHRNA3 LNCR2,
NACHRA3,
PAOD2
1.8kb Sense Chr. 15 - 78593052 - 78621295 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010150.hg.1 ENST00000567141.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010150.hg.1 ENST00000567141.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0