This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03675243_s1
See other ALPK2 GE Assays ›
Gene Symbol:
ALPK2
Gene Name:
alpha kinase 2
Gene Aliases:
HAK
Chromosome Location:
Chr.18: 58481247 - 58629192 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03675243_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALPK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK125243.1 - 1 1647 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 122 MIR122 MIR122A,
MIRN122,
MIRN122A,
hsa-mir-122,
miRNA122,
miRNA122A,
mir-122
-126.8kb Anti-sense Chr. 18 - 58451074 - 58451158 on NCBI Build GRCh38
Downstream uncharacterized LOC102724745 LOC102724745 78.4kb Anti-sense Chr. 18 - 58658510 - 58669343 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590642.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000361673.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587399.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000587842.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000589204.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590642.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 ENST00000590662.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq
Affymetrix Human Clariom D Assay TC1800008804.hg.1 NM_052947.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0