This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03674655_s1
See other CD4 GE Assays ›
Gene Symbol:
CD4
Gene Name:
CD4 molecule
Gene Aliases:
CD4mut
Chromosome Location:
Chr.12: 6789472 - 6820810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03674655_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA S79267.1 - 1 1545 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream lymphocyte activating 3 LAG3 CD223 -20.5kb Sense Chr. 12 - 6772483 - 6778455 on NCBI Build GRCh38
Downstream G protein-coupled receptor 162 GPR162 A-2,
GRCA
20.7kb Sense Chr. 12 - 6821797 - 6827418 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000011653.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000437800.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000535707.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536563.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000536610.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000538827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000539492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000541982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000543755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000543755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000543755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000543755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000543755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 ENST00000544344.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_000616.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195014.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195015.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195016.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200006645.hg.1 NM_001195017.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0