This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03674179_s1
See other CD47 GE Assays ›
Gene Symbol:
CD47
Gene Name:
CD47 molecule
Gene Aliases:
IAP, MER6, OA3
Chromosome Location:
Chr.3: 108043094 - 108094200 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03674179_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD47
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC042889.1 - 1 2649 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY994485.1 - 1 Chr. 3 - 108047079 - 108047614 [+] on Build GRCh38 111
GenBankmRNA DB350957.1 - 1 Chr. 3 - 108047062 - 108047609 [+] on Build GRCh38 111
GenBankmRNA BQ002786.1 - 1 Chr. 3 - 108047078 - 108047822 [+] on Build GRCh38 111
GenBankmRNA BM983263.1 - 1 587 Chr. 3 - 108047376 - 108048073 [-] on Build GRCh38 111
GenBankmRNA BM697721.1 - 1 353 Chr. 3 - 108047230 - 108047839 [-] on Build GRCh38 111
GenBankmRNA AW166867.1 - 1 Chr. 3 - 108047079 - 108047571 [+] on Build GRCh38 111
GenBankmRNA AL700103.1 - 1 399 Chr. 3 - 108047057 - 108047885 [-] on Build GRCh38 111
GenBankmRNA BM151464.1 - 1 Chr. 3 - 108047079 - 108047567 [+] on Build GRCh38 111
GenBankmRNA BG230614.1 - 1 Chr. 3 - 108047079 - 108047621 [+] on Build GRCh38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 636 LINC00636 -118.2kb Anti-sense Chr. 3 - 107883205 - 107928907 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1215 LINC01215 74.9kb Anti-sense Chr. 3 - 108125064 - 108138610 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0