Assay Details
Gene Symbol: DACOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY203937.1 | - | 2 | 1153 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102723493 | LOC102723493 | Sense | Chr. 15 - 66984207 - 67064182 on NCBI Build GRCh38 | ||
Downstream | SMAD family member 3 | SMAD3 |
HSPC193, HsT17436, JV15-2, LDS1C, LDS3, MADH3 |
20.4kb | Anti-sense | Chr. 15 - 67065698 - 67195195 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726528 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726505 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726544 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726568 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219687 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219657 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726478 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799846 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799835 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799834 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726506 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726502 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219628 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219685 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726517 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726514 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09726500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16799841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30219643 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 279024 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 762129 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 849334 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 93856 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3297883 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13991 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20287136 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33734447 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P33734457 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20287140 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P33734447 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P33734457 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20287136 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | XM_001724622_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XM_001725451_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XM_001726695_at | - | - |
Affymetrix Human Gene 1.1 ST Array | 7989951 | - | - |
