This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673766_s1
See other LOC286058 GE Assays ›
Gene Symbol:
LOC286058
Gene Name:
uncharacterized LOC286058
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673766_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC286058
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL833160.1 - 1 4648 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream bridging integrator 3 BIN3 -9.9kb Sense Chr. 8 - 22620418 - 22669148 on NCBI Build GRCh38
Downstream early growth response 3 EGR3 EGR-3,
PILOT
3.7kb Sense Chr. 8 - 22687661 - 22693302 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 210505 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 469605 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 211296 BC118576.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 211296 BC118657.1 genbank_mrna_human
Agilent SurePrint G3 Human CNV Microarray 2x400K 211296 NM_001077624.1 refseq_mrna_human_NM
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3630780 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56339 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3630780 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P38334392 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38334392 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01878229 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P25962209 - -
Agilent Human 1B Microarray G4111A A_32_P363217 - -
Agilent Human 1B Microarray G4111A A_32_P393682 - -
Agilent Human 1B Microarray G4111A A_32_P348007 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38334392 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1562247_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009855.hg.1 ENST00000566457.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009855.hg.1 ENST00000566457.1 Ensembl
Human Genome U133 Plus 2.0 Array 1562247_PM_at - -
Human Genome U133 Plus 2.0 Array 1562247_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1562247_at4 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0