This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673504_s1
See other LOC100133032 GE Assays ›
Gene Symbol:
LOC100133032
Gene Name:
uncharacterized LOC100133032
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673504_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100133032
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK130228.1 - 1 246 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chondroitin sulfate proteoglycan 5 CSPG5 NGC -23.1kb Sense Chr. 3 - 47562238 - 47580240 on NCBI Build GRCh38
Downstream DEAH-box helicase 30 DHX30 DDX30,
RETCOR
198.2kb Anti-sense Chr. 3 - 47802909 - 47850196 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK130228.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3326892 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55857 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300010992.hg.1 ENST00000398957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010992.hg.1 ENST00000398957.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010992.hg.1 ENST00000398957.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010992.hg.1 ENST00000398957.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0