This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03673275_s1
See other LOC100128281 GE Assays ›
Gene Symbol:
LOC100128281
Gene Name:
uncharacterized LOC100128281
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03673275_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100128281
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK128353.1 - 2 1755 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX117344.1 - 1 306 Chr. 17 - 7868684 - 7869217 [-] on Build GRCh38 98
GenBankmRNA AI219813.1 - 1 Chr. 17 - 7868689 - 7869110 [+] on Build GRCh38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cytochrome b5 domain containing 1 CYB5D1 -4.3kb Anti-sense Chr. 17 - 7857746 - 7864417 on NCBI Build GRCh38
Downstream chromodomain helicase DNA binding protein 3 CHD3 Mi-2a,
Mi2-ALPHA,
ZFH
15.2kb Anti-sense Chr. 17 - 7884764 - 7912757 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3243307 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21097 - -
Agilent Human Genome CGH + SNP Microarray A_16_P20578956 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3243307 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1565723_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0