This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03671229_s1
See other RCBTB2 GE Assays ›
Gene Symbol:
RCBTB2
Gene Name:
RCC1 and BTB domain containing protein 2
Gene Aliases:
CHC1L, RLG
Chromosome Location:
Chr.13: 48488963 - 48535995 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03671229_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RCBTB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC031965.1 - 1 542 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RB transcriptional corepressor 1 RB1 OSRC,
PPP1R130,
RB,
p105-Rb,
pRb,
pp110
-51.0kb Sense Chr. 13 - 48303747 - 48481890 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 462 LINC00462 43.2kb Anti-sense Chr. 13 - 48576974 - 48580901 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0