This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03669519_s1
See other CD36 GE Assays ›
Gene Symbol:
CD36
Gene Name:
CD36 molecule
Gene Aliases:
BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
Chromosome Location:
Chr.7: 80602188 - 80679277 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03669519_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057690.1 - 1 2050 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein subunit alpha transducin 3 GNAT3 GDCA -134.2kb Anti-sense Chr. 7 - 80457291 - 80512503 on NCBI Build GRCh38
Downstream semaphorin 3C SEMA3C SEMAE,
SemE
77.2kb Anti-sense Chr. 7 - 80742538 - 80919351 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700011625.hg.1 ENST00000449243.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011625.hg.1 ENST00000449243.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 207290 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0