This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03668459_s1
See other GUSBP11 GE Assays ›
Gene Symbol:
GUSBP11
Gene Name:
glucuronidase, beta pseudogene 11
Gene Aliases:
-
Chromosome Location:
Chr.22: 23638487 - 23717423 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03668459_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GUSBP11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK025313.1 - 1 2649 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream aspartate rich 1 DRICH1 C22orf43,
KB-208E9.1
-43.1kb Sense Chr. 22 - 23580872 - 23637732 on NCBI Build GRCh38
Downstream ral guanine nucleotide dissociation stimulator like 4 RGL4 Rgr 6.8kb Anti-sense Chr. 22 - 23690861 - 23699176 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0