This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03665226_s1
See other ASH1L-AS1 GE Assays ›
Gene Symbol:
ASH1L-AS1
Gene Name:
ASH1L antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 155561981 - 155563944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03665226_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASH1L-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC065229.1 - 1 854 120
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM686576.1 - 1 304 Chr. 1 - 155563818 - 155564348 [+] on Build GRCh38 120

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ASH1 like histone lysine methyltransferase ASH1L ASH1,
ASH1L1,
KMT2H
-410.0 Anti-sense Chr. 1 - 155335261 - 155563160 on NCBI Build GRCh38
Downstream misato 1,
mitochondrial distribution and morphology regulator
MSTO1 LST005,
MST
45.8kb Sense Chr. 1 - 155610170 - 155614967 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000597216.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009949.hg.1 ENST00000599360.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000452809.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000456633.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 ENST00000594351.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq
Affymetrix Human Clariom D Assay TC0100010129.hg.1 NR_027023.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0