This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03663886_m1
See other LINC01344 GE Assays ›
Gene Symbol:
LINC01344
Gene Name:
long intergenic non-protein coding RNA 1344
Gene Aliases:
GS1-122H1.2
Chromosome Location:
Chr.1: 182203945 - 182314061 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03663886_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01344
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK128015.1 - 9 - 10 1807 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger protein 648 ZNF648 -91.2kb Sense Chr. 1 - 182053740 - 182205330 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000420760.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000449842.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 ENST00000608183.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Clariom D Assay TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Clariom D Assay TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Clariom D Assay TC0100016621.hg.1 NR_104175.1 Refseq
Affymetrix Human Clariom D Assay TC0100016621.hg.1 NR_104175.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0