This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03663620_s1
See other LOC401433 GE Assays ›
Gene Symbol:
LOC401433 WDR86
Gene Name:
uncharacterized LOC401433
WD repeat domain 86
Gene Aliases:
-
-
Chromosome Location:
-
Chromosome Location:
Chr.7: 151377974 - 151410705 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs03663620_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC401433
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK127717.1 - 1 1951 102
Gene Symbol: WDR86
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011516144.2 XP_011514446.1 7 3658 102
XM_011516145.2 XP_011514447.1 6 3529 102
XM_011516147.2 XP_011514449.1 6 3458 102
XM_011516151.2 XP_011514453.1 5 3329 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream negative regulator of ubiquitin like proteins 1 NUB1 BS4,
NUB1L,
NYREN18
Anti-sense Chr. 7 - 151341699 - 151378461 on NCBI Build GRCh38
Downstream WDR86 antisense RNA 1 WDR86-AS1 28.7kb Anti-sense Chr. 7 - 151409161 - 151413354 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3831099 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20887 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3831099 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18166569 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01835962 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array XR_017807_at - -
Affymetrix Human Gene 1.0 ST Array XR_040276_at - -
Affymetrix Human Gene 1.0 ST Array XR_017805_at - -
Affymetrix Human Gene 1.0 ST Array XR_017935_at - -
Affymetrix Human Gene 1.0 ST Array 8143940 - -
Affymetrix Human Gene 1.1 ST Array 8143939 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3279640 AK127717.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3207086 - -
HumanHT-12 V4.0 expression beadchip ILMN_3207086 - -
HumanHT-12 V4.0 expression beadchip ILMN_3279640 AK127717.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1766386 - -
Illumina humanRef-8 v2.0 expression beadchip ILMN_1766386 - -
Illumina HumanWG-6_V2_0_R2 ILMN_1912433 - -
Illumina HumanWG-6_V2_0_R2 ILMN_1766386 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0