This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03660898_s1
See other LOC100129846 GE Assays ›
Gene Symbol:
LOC100129846
Gene Name:
uncharacterized LOC100129846
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03660898_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100129846
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK124859.1 - 1 3849 84
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA100546.1 - 1 Chr. 8 - 30570371 - 30570757 [-] on Build GRCh38 84
GenBankmRNA AW044139.1 - 1 Chr. 8 - 30570414 - 30570773 [-] on Build GRCh38 84
GenBankmRNA DB308234.1 - 1 Chr. 8 - 30570212 - 30570756 [-] on Build GRCh38 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RBPMS antisense RNA 1 RBPMS-AS1 -181.4kb Anti-sense Chr. 8 - 30382119 - 30385401 on NCBI Build GRCh38
Downstream general transcription factor IIE subunit 2 GTF2E2 FE,
TF2E2,
TFIIE-B,
TTD6
7.8kb Anti-sense Chr. 8 - 30578514 - 30658241 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3337277 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37387 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3337277 - -
Array Name Probe IDs Targets Target Type
Human Genome U95 Set 78856_at - -
Human Genome U95 Set 78856_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0