This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03659122_s1
See other CEBPB-AS1 GE Assays ›
Gene Symbol:
CEBPB-AS1
Gene Name:
CEBPB antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 50184603 - 50192069 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03659122_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPB-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125739.1 7 1274 67
GenBank mRNA AK129842.1 - 1 49 67
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX452605.2 - 2 166 Chr. 20 - 50185996 - 50187057 [-] on Build GRCh38 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 1273 LINC01273 -7.9kb Anti-sense Chr. 20 - 50172570 - 50176671 on NCBI Build GRCh38
Downstream CCAAT/enhancer binding protein beta CEBPB C/EBP-beta,
IL6DBP,
NF-IL6,
TCF5
4.2kb Anti-sense Chr. 20 - 50190583 - 50192690 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P31853862 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204652 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204653 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204643 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204648 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P31853870 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P31853857 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204639 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204647 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P31853867 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204654 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204644 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P11204641 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 57208 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3781228 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21643 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3781228 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41348468 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0