This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03657907_s1
See other LOC729887 GE Assays ›
Gene Symbol:
LOC729887
Gene Name:
uncharacterized LOC729887
Gene Aliases:
-
Chromosome Location:
Chr.16: 84116181 - 84117571 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03657907_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC729887
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC052334.1 - 1 454 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream membrane bound transcription factor peptidase,
site 1
MBTPS1 PCSK8,
S1P,
SKI-1
-108.0 Anti-sense Chr. 16 - 84053761 - 84116943 on NCBI Build GRCh38
Downstream hydroxysteroid dehydrogenase like 1 HSDL1 SDR12C3 4.6kb Anti-sense Chr. 16 - 84122139 - 84145195 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_32_P101699 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P101699 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 31149 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58520 - -
SurePrint G3 Human GE Microarray G4851A A_32_P101699 - -
Whole Human Genome Oligo Microarray G4112A A_32_P101699 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P101699 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 230591_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC1600008636.hg.1 ENST00000565382.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008636.hg.1 ENST00000565382.1 Ensembl
Human Genome U133 Plus 2.0 Array 230591_at - -
Human Genome U133 Plus 2.0 Array 230591_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 230591_at5 - -
Human Genome U95 Set 76590_at - -
Human Genome U95 Set 76590_PM_at - -
Human Genome U133 Set 230591_PM_at - -
Human Genome U133 Set 230591_at - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3221760 - -
HumanHT-12 v4 Expression BeadChip ILMN_3301740 BC052334.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3301740 BC052334.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_3221760 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0