Assay Details
Gene Symbol: COPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC037941.2 | - | 1 | 147 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SUMO1 pseudogene 3 | SUMO1P3 | -6.0kb | Anti-sense | Chr. 1 - 160317265 - 160318474 on NCBI Build GRCh38 | |
Downstream | nicastrin | NCSTN | ATAG1874 | 17.9kb | Anti-sense | Chr. 1 - 160343056 - 160358952 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
