This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03657176_s1
See other XPNPEP3 GE Assays ›
Gene Symbol:
XPNPEP3
Gene Name:
X-prolyl aminopeptidase 3
Gene Aliases:
APP3, ICP55, NPHPL1
Chromosome Location:
Chr.22: 40857081 - 40932819 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03657176_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: XPNPEP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC037424.2 - 1 753 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW593748.1 - 1 Chr. 22 - 40857764 - 40858025 [-] on Build GRCh38 94
GenBankmRNA BE617907.1 - 1 Chr. 22 - 40857190 - 40857978 [-] on Build GRCh38 94
GenBankmRNA AI127653.1 - 1 Chr. 22 - 40857551 - 40858031 [-] on Build GRCh38 94
GenBankmRNA AW002166.1 - 1 Chr. 22 - 40857620 - 40858028 [-] on Build GRCh38 94
GenBankmRNA AI471784.1 - 1 Chr. 22 - 40857556 - 40858025 [-] on Build GRCh38 94
GenBankmRNA AW005574.1 - 1 Chr. 22 - 40857501 - 40858026 [-] on Build GRCh38 94
GenBankmRNA CB409437.1 - 1 421 Chr. 22 - 40857423 - 40858004 [+] on Build GRCh38 94
GenBankmRNA DB546196.1 - 1 147 Chr. 22 - 40857697 - 40858025 [+] on Build GRCh38 94
GenBankmRNA CB409178.1 - 1 421 Chr. 22 - 40857423 - 40858013 [+] on Build GRCh38 94
GenBankmRNA BE674036.1 - 1 Chr. 22 - 40857435 - 40858030 [-] on Build GRCh38 94
GenBankmRNA BM466568.1 - 1 633 Chr. 22 - 40857233 - 40858036 [+] on Build GRCh38 94
GenBankmRNA AA398384.1 - 1 Chr. 22 - 40857639 - 40858026 [-] on Build GRCh38 94
GenBankmRNA CA314208.1 - 1 Chr. 22 - 40857341 - 40858031 [-] on Build GRCh38 94
GenBankmRNA AI652199.1 - 1 Chr. 22 - 40857632 - 40858025 [-] on Build GRCh38 94
GenBankmRNA AI885037.1 - 1 Chr. 22 - 40857523 - 40858031 [-] on Build GRCh38 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) ST13 AAG2,
FAM10A1,
FAM10A4,
HIP,
HOP,
HSPABP,
HSPABP1,
P48,
PRO0786,
SNC6
-83.0 Anti-sense Chr. 22 - 40824535 - 40857008 on NCBI Build GRCh38
Downstream DnaJ heat shock protein family (Hsp40) member B7 DNAJB7 DJ5,
HSC3
1.5kb Anti-sense Chr. 22 - 40859549 - 40862126 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0