This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03655180_s1
See other HAMP GE Assays ›
Gene Symbol:
HAMP
Gene Name:
hepcidin antimicrobial peptide
Gene Aliases:
HEPC, HFE2B, LEAP1, PLTR
Chromosome Location:
Chr.19: 35282346 - 35285143 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03655180_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK096110.1 - 1 2155 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB321269.1 - 1 Chr. 19 - 35284609 - 35285140 [-] on Build GRCh38 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream upstream transcription factor 2,
c-fos interacting
USF2 FIP,
bHLHb12
-2.7kb Sense Chr. 19 - 35268978 - 35279821 on NCBI Build GRCh38
Downstream myelin associated glycoprotein MAG GMA,
S-MAG,
SIGLEC-4A,
SIGLEC4A,
SPG75
6.9kb Sense Chr. 19 - 35292086 - 35313807 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000222304.3 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000593580.1 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 ENST00000598398.5 Ensembl
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq
Affymetrix Human Clariom D Assay TC1900007848.hg.1 NM_021175.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0