This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03654155_m1
See other MKNK1 GE Assays ›
Gene Symbol:
MKNK1
Gene Name:
MAP kinase interacting serine/threonine kinase 1
Gene Aliases:
MNK1
Chromosome Location:
Chr.1: 46557407 - 46607140 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03654155_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MKNK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024174.2 5 - 6 556 59
NR_024176.2 6 - 7 657 59
XM_006711013.1 XP_006711076.1 5 - 6 508 59
XM_006711014.1 XP_006711077.1 5 - 6 499 59
XM_006711015.1 XP_006711078.1 6 - 7 570 59
XM_017002664.1 XP_016858153.1 5 - 6 5984 59
XM_017002666.1 XP_016858155.1 6 - 7 3459 59
XM_017002667.1 XP_016858156.1 4 - 5 1035 59
GenBank mRNA AK298839.1 - 5 - 6 530 59
BX538193.1 - 4 - 5 380 59
DC366212.1 - 5 - 6 530 59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DC407161.1 - 5 - 6 513 Chr. 1 - 46572097 - 46604251 [-] on Build GRCh38 59
GenBankmRNA DC387738.1 - 5 - 6 486 Chr. 1 - 46572090 - 46604266 [-] on Build GRCh38 59
GenBankmRNA DC371403.1 - 5 - 6 488 Chr. 1 - 46572089 - 46604268 [-] on Build GRCh38 59
GenBankmRNA DA998146.1 - 3 - 4 314 Chr. 1 - 46565056 - 46604252 [-] on Build GRCh38 59
GenBankmRNA DC405883.1 - 5 - 6 508 Chr. 1 - 46572120 - 46604246 [-] on Build GRCh38 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MKNK1 antisense RNA 1 MKNK1-AS1 -1.8kb Anti-sense Chr. 1 - 46538696 - 46570255 on NCBI Build GRCh38
Downstream MOB kinase activator 3C MOB3C MOB1E,
MOBKL2C
31.1kb Sense Chr. 1 - 46607715 - 46616891 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of translational initiation
protein phosphorylation
peptidyl-serine phosphorylation
intracellular signal transduction
protein autophosphorylation
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0