This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03461324_m1
See other ZFAS1 GE Assays ›
Gene Symbol:
ZFAS1
Gene Name:
ZNFX1 antisense RNA 1
Gene Aliases:
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1
Chromosome Location:
Chr.20: 49278178 - 49289260 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03461324_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZFAS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003605.1 1 - 2 224 106
GenBank mRNA AY513723.1 - 1 - 2 224 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA771146.1 - 4 - 5 Chr. 20 - 49278274 - 49289258 [-] on Build GRCh38 106
GenBankmRNA CA771453.1 - 1 - 2 131 Chr. 20 - 49278271 - 49289111 [+] on Build GRCh38 106
GenBankmRNA BU076692.1 - 1 - 2 131 Chr. 20 - 49278271 - 49289260 [+] on Build GRCh38 106
GenBankmRNA BU076411.1 - 4 - 5 Chr. 20 - 49278253 - 49289258 [-] on Build GRCh38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream zinc finger NFX1-type containing 1 ZNFX1 Anti-sense Chr. 20 - 49245897 - 49278244 on NCBI Build GRCh38
Downstream small nucleolar RNA,
C/D box 12B
SNORD12B 1.1kb Sense Chr. 20 - 49280313 - 49280415 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000009932.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000386.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 NR_003604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 NR_003605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 NR_003606.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 NR_036658.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005618.hg.1 TC20000386.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 NR_003604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 NR_003605.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 NR_003606.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 NR_036658.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005619.hg.1 TC20000386.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170073.hg.1 TC2000009932.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170074.hg.1 TC2000009932.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000326677.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000326677.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000428008.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000428008.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000610973.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_036659.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000326677.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000326677.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000371743.3 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000417721.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000428008.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000428008.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000441722.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000450535.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000458653.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 ENST00000610973.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003604.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003605.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_003606.3 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_036658.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 NR_036659.2 Refseq
Affymetrix Human Clariom D Assay PSR2000170087.hg.1 TC2000009932.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0