This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03454934_m1
See other RMST GE Assays ›
Gene Symbol:
RMST
Gene Name:
rhabdomyosarcoma 2 associated transcript (non-protein coding)
Gene Aliases:
LINC00054, NCRMS, NCRNA00054
Chromosome Location:
Chr.12: 97465021 - 97533766 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03454934_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RMST
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF429306.1 - 5 - 6 906 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 135a-2 MIR135A2 MIRN135-2,
MIRN135A2,
mir-135a-2
Sense Chr. 12 - 97563812 - 97563911 on NCBI Build GRCh38
Downstream platelet activating factor acetylhydrolase 1b catalytic subunit 2 pseudogene LOC643711 149.1kb Anti-sense Chr. 12 - 97713412 - 97756517 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000610287.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000611651.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000611980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000613072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000616309.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000616886.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000617263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000618704.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000619945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 ENST00000621935.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000538559.2 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000541282.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547946.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000547996.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000548886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000552411.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000610287.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000611651.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000611980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000613072.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000616309.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000616886.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000617263.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000618704.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000619945.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 ENST00000621935.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq
Affymetrix Human Clariom D Assay TC1200008551.hg.1 NR_024037.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0