This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03454515_m1
See other LOC101927730 GE Assays ›
Gene Symbol:
LOC101927730
Gene Name:
uncharacterized LOC101927730
Gene Aliases:
-
Chromosome Location:
Chr.6: 2883988 - 2900676 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03454515_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927730
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110841.1
3 - 4 317 107
GenBank mRNA AY927511.1 - LIT3521
AY927511
2 - 3 174 107
AY927512.1 - LIT3522
AY927512
3 - 4 317 107

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream serpin family B member 9 pseudogene 1 SERPINB9P1 SERPINB9P -22.5kb Anti-sense Chr. 6 - 2854657 - 2876510 on NCBI Build GRCh38
Downstream serpin family B member 6 SERPINB6 CAP,
DFNB91,
MSTP057,
PI-6,
PI6,
PTI,
SPI3
47.5kb Anti-sense Chr. 6 - 2948159 - 2972165 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header