This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03454344_m1
See other DLEU1 GE Assays ›
Gene Symbol:
DLEU1
Gene Name:
deleted in lymphocytic leukemia 1 (non-protein coding)
Gene Aliases:
BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6
Chromosome Location:
Chr.13: 50082169 - 50528643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03454344_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLEU1
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109974.1
5 - 6 789 115
GenBank mRNA AJ412028.1 - LIT1924
BCMS
3 - 4 322 115
AJ412055.1 - LIT1885
BCMS
5 - 6 787 115
AJ412059.1 - LIT1874
BCMS
6 - 7 900 115
AJ412060.1 - LIT1873
BCMS
4 - 5 1886 115

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DLEU1 antisense RNA 1 DLEU1-AS1 LINC01308 Anti-sense Chr. 13 - 50520933 - 50527449 on NCBI Build GRCh38
Downstream deleted in lymphocytic leukemia,
7
DLEU7 167.5kb Anti-sense Chr. 13 - 50711026 - 50843939 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0