This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03406429_gH
See other IFNA10 GE Assays ›
Gene Symbol:
IFNA10 IFNA4
Gene Name:
interferon alpha 10
interferon alpha 4
Gene Aliases:
IFN-alphaC
IFN-alpha4a, INFA4
Chromosome Location:
Chr.9: 21206181 - 21207143 on Build GRCh38
Chromosome Location:
Chr.9: 21186618 - 21187599 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs03406429_gH
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFNA10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002171.2 NP_002162.1 1 55 130
GenBank mRNA BC069409.1 - 1 55 7262186 130
BC103972.2 - 1 55 40015081 130
BC103973.1 - 1 55 40015082 130
V00551.1 - 1 55 130
Gene Symbol: IFNA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021068.2 NP_066546.1 1 77 130
GenBank mRNA BC113640.1 - 1 65 8322692 130
BC113642.1 - 1 65 8322694 130
M27318.1 - 1 149 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009694.hg.1, TC0900012214.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000950.hg.1, TC09000952.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC069409.1,BC103972.2,BC103973.1,BC113640.1,BC113642.1,M27318.1,NM_002171.2,NM_021068.2,V00551.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFNA10
Click here to find siRNAs for Gene IFNA4

Panther Classification:

Molecular Function -

interferon superfamily

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
humoral immune response
blood coagulation
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
innate immune response
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
response to virus
cytokine activity
type I interferon receptor binding
protein binding

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P16164884 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26925806 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06637773 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P26925807 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P06637772 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 39979 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 209100 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P20704 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P20704 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 9165 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43532 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P20704 - -
SurePrint G3 Human GE Microarray G4851A A_23_P20704 - -
SurePrint G3 Human GE Microarray G4851A ERCC-00123_60 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925806 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637769 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637772 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637773 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16164884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925809 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16164884 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637773 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637772 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925809 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925807 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925792 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26925806 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06637769 - -
Whole Human Genome Oligo Microarray G4112A A_23_P20704 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06637770 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06637769 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06637772 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P06637771 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P20704 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06637771 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06637767 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06637766 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16164847 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P06637772 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_002171_at - -
Affymetrix Human Gene 1.0 ST Array 8160379 - -
Affymetrix Human Gene 1.0 ST Array 8160381 - -
Affymetrix Human Gene 1.0 ST Array 8160397 - -
Affymetrix Human Gene 1.0 ST Array 8160395 - -
Affymetrix Human Gene 1.0 ST Array 8160398 - -
Affymetrix Human Gene 1.0 ST Array 8160390 - -
Affymetrix Human Gene 1.0 ST Array 8160380 - -
Affymetrix Human Gene 1.0 ST Array 8160396 - -
Affymetrix Human Gene 1.0 ST Array 8160391 - -
Affymetrix Human Gene 1.0 ST Array 8160378 - -
Affymetrix Human Gene 1.1 ST Array 8160377 - -
Affymetrix Human Gene 1.1 ST Array 8160389 - -
Affymetrix Human Gene 1.1 ST Array 8160394 - -
Affymetrix HT HG-U133+ PM Array Plate 208261_PM_x_at - -
Affymetrix Human Genome U219 Array 11715139_PM_x_at - -
Affymetrix Human Genome U219 Array 11715139_x_at - -
Affymetrix Human Transcriptome Array 2.0 TC0900012215.hg.1 ENST00000380216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012215.hg.1 ENST00000380216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012215.hg.1 NM_002173.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012215.hg.1 NM_002173.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012679.hg.1 ENST00000421715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012679.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012679.hg.1 TC09000950.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012697.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012697.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012697.hg.1 TC09000952.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012695.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012695.hg.1 NM_002171.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012695.hg.1 TC09000952.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09012680.hg.1 ENST00000421715.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012680.hg.1 NM_021068.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012680.hg.1 TC09000950.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900185707.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185707.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185707.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185707.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185707.hg.1 TC0900012214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900185710.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185710.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185710.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185710.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185710.hg.1 TC0900012214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900185711.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185711.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185711.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185711.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185711.hg.1 TC0900012214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900009694.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay TC0900012214.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay TC0900012214.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay TC0900012215.hg.1 ENST00000380216.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012215.hg.1 ENST00000380216.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012215.hg.1 NM_002173.3 Refseq
Affymetrix Human Clariom D Assay TC0900012215.hg.1 NM_002173.3 Refseq
Affymetrix Human Clariom D Assay PSR0900185709.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185709.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185709.hg.1 ENST00000357374.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900185709.hg.1 NM_002171.2 Refseq
Affymetrix Human Clariom D Assay PSR0900185709.hg.1 TC0900012214.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900166323.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166323.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay PSR0900166323.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166323.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay PSR0900166323.hg.1 TC0900009694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900166325.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166325.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay PSR0900166325.hg.1 ENST00000421715.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166325.hg.1 NM_021068.2 Refseq
Affymetrix Human Clariom D Assay PSR0900166325.hg.1 TC0900009694.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 208261_x_at - -
Human Genome U133 Plus 2.0 Array 208261_PM_x_at - -
Human Genome U133A 2.0 Array 208261_x_at - -
Human Genome U133A 2.0 Array 208261_PM_x_at - -
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_002171_at - -
AffymetrixPathwork Diagnostics Pathchip pw574397 - -
Human Genome U95 Set 427_PM_f_at - -
Human Genome U95 Set 427_f_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 208261_x_at4 - -
Human Genome U133 Set 208261_PM_x_at - -
Human Genome U133 Set 208261_x_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A037177 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A037177 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 208360 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1769988 V00551.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_4504588-S BC103972.2 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_4504588-S BC103973.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_4504588-S V00551.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_4504588-S NM_002171.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_4504588-S BC069409.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_4745-S - -
Illumina human-6 v1.0 expression beadchip 0000450463 - -
Illumina humanRef-8 v1.0 expression beadchip 0000450463 - -
Illumina human-6 v2.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip 2690367 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_4745 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1769988 NM_002171.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1769988 V00551.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1769988 NM_002171.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0