This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03296892_s1
See other TMEM185B GE Assays ›
Gene Symbol:
TMEM185B
Gene Name:
transmembrane protein 185B
Gene Aliases:
FAM11B
Chromosome Location:
Chr.2: 120217471 - 120223408 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03296892_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM185B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024121.2 NP_077026.2 1 2050 133
GenBank mRNA AF530474.1 - 1 2050 133
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA337541.1 - 1 425 Chr. 2 - 120221209 - 120221782 [-] on Build GRCh38 133
GenBankmRNA DB277964.1 - 1 267 Chr. 2 - 120221057 - 120221624 [-] on Build GRCh38 133
GenBankmRNA CA426885.1 - 1 Chr. 2 - 120221268 - 120221841 [+] on Build GRCh38 133
GenBankmRNA AA706579.1 - 1 Chr. 2 - 120221268 - 120221652 [+] on Build GRCh38 133
GenBankmRNA CA306194.1 - 1 Chr. 2 - 120221280 - 120221944 [+] on Build GRCh38 133
GenBankmRNA AA759372.1 - 1 Chr. 2 - 120221280 - 120221657 [+] on Build GRCh38 133
GenBankmRNA DA221735.1 - 1 132 Chr. 2 - 120220933 - 120221490 [-] on Build GRCh38 133
GenBankmRNA CN373477.1 - 1 280 Chr. 2 - 120221120 - 120221630 [-] on Build GRCh38 133
GenBankmRNA CN373479.1 - 1 111 Chr. 2 - 120220983 - 120221469 [-] on Build GRCh38 133
GenBankmRNA CR743649.1 - 1 288 Chr. 2 - 120221184 - 120221645 [-] on Build GRCh38 133
GenBankmRNA AA844074.1 - 1 Chr. 2 - 120221274 - 120221659 [+] on Build GRCh38 133
GenBankmRNA DA116246.1 - 1 425 Chr. 2 - 120221205 - 120221783 [-] on Build GRCh38 133

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream erythrocyte membrane protein band 4.1 like 5 EPB41L5 BE37,
YMO1,
YRT
-38.4kb Anti-sense Chr. 2 - 120013028 - 120179121 on NCBI Build GRCh38
Downstream RALB Ras like proto-oncogene B RALB 16.7kb Anti-sense Chr. 2 - 120240070 - 120294711 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014049.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002256.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02036353.hg.1 NM_024121.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036353.hg.1 TC02002256.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200202847.hg.1 NM_024121.2 Refseq
Affymetrix Human Clariom D Assay PSR0200202847.hg.1 NM_024121.2 Refseq
Affymetrix Human Clariom D Assay PSR0200202847.hg.1 TC0200014049.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0