This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03044517_m1
See other FAM71F1 GE Assays ›
Gene Symbol:
FAM71F1
Gene Name:
family with sequence similarity 71 member F1
Gene Aliases:
FAM137A, NYD-SP18
Chromosome Location:
Chr.7: 128709061 - 128731743 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03044517_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM71F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282788.1 NP_001269717.1 2 - 3 529 135
NM_032599.3 NP_115988.1 2 - 3 529 135
NR_104242.1 2 - 3 529 135
XM_017012743.1 XP_016868232.1 2 - 3 572 135
GenBank mRNA AF367470.1 - 2 - 3 482 135
AK310308.1 - 2 - 3 476 135
AK310426.1 - 3 - 4 385 135
AK313608.1 - 2 - 3 480 135
BC037248.1 - 2 - 3 468 5165912 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000781.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAM71F1

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07012283.hg.1 ENST00000315184.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07012283.hg.1 NM_032599.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07012283.hg.1 TC07000781.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header