This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03025927_s1
See other GDPGP1 GE Assays ›
Gene Symbol:
GDPGP1
Gene Name:
GDP-D-glucose phosphorylase 1
Gene Aliases:
C15orf58, VTC2
Chromosome Location:
Chr.15: 90234255 - 90242080 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs03025927_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDPGP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013657.2 NP_001013679.2 4 1155 98
NM_001322811.1 NP_001309740.1 5 1274 98
GenBank mRNA AK130527.1 - 4 1741 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500008304.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15000859.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GDPGP1

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
positive regulation of GTPase activity
nucleotide binding
guanyl-nucleotide exchange factor activity
nucleotidyltransferase activity
hydrolase activity
GDP-D-glucose phosphorylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 ENST00000565730.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15008020.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15008020.hg.1 TC15000859.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15008021.hg.1 ENST00000558017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15008021.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15008021.hg.1 TC15000859.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15008024.hg.1 ENST00000329600.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15008024.hg.1 ENST00000558017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15008024.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15008024.hg.1 TC15000859.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR15008022.hg.1 ENST00000558017.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15008022.hg.1 NM_001013657.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15008022.hg.1 TC15000859.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159893.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159885.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159887.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159894.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 ENST00000565730.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay TC1500008304.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159889.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000558291.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000559204.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 ENST00000561433.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159892.hg.1 TC1500008304.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 ENST00000329600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 ENST00000558017.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 NM_001013657.2 Refseq
Affymetrix Human Clariom D Assay PSR1500159895.hg.1 TC1500008304.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0