This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02871145_g1
See other GOLGA7 GE Assays ›
Gene Symbol:
GOLGA7
Gene Name:
golgin A7
Gene Aliases:
GCP16, GOLGA3AP1, GOLGA7A, HSPC041
Chromosome Location:
Chr.8: 41490562 - 41510980 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02871145_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOLGA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001174124.1 NP_001167595.1 1 - 2 62 59
NM_016099.2 NP_057183.2 1 - 2 62 59
GenBank mRNA AB104615.1 - 1 - 2 42 59
AF125102.1 - 1 - 2 28 59
AK026613.1 - 1 - 2 42 59
AL695304.1 - 1 - 2 71 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB104615.1, AF125102.1, NM_001174124.1, NM_016099.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GOLGA7

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
peptidyl-L-cysteine S-palmitoylation
Golgi to plasma membrane protein transport
protein stabilization
protein binding
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0