This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02789702_m1
See other GNRH1 GE Assays ›
Gene Symbol:
GNRH1
Gene Name:
gonadotropin releasing hormone 1
Gene Aliases:
GNRH, GRH, LHRH, LNRH
Chromosome Location:
Chr.8: 25419258 - 25425040 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02789702_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNRH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000825.3 NP_000816.4 1 - 2 1846 71
NM_001083111.1 NP_001076580.1 2 - 3 977 71
GenBank mRNA BC126437.1 - 1 - 2 668 8992153 71
BC126463.1 - 1 - 2 667 8992179 71
CB243067.1 - 2 - 3 321 71
M12578.1 - 2 - 3 168 71
X01059.1 - 1 - 2 1210 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GNRH1

Panther Classification:

Molecular Function -

peptide hormone intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
cell-cell signaling
multicellular organism development
female pregnancy
aging
negative regulation of cell proliferation
regulation of gene expression
male sex determination
response to corticosteroid
response to lipopolysaccharide
negative regulation of immature T cell proliferation
response to testosterone
response to prostaglandin E
response to potassium ion
negative regulation of apoptotic process
estrous cycle
response to ethanol
response to prolactin
regulation of ovarian follicle development
negative regulation of neuron migration
hormone activity
gonadotropin hormone-releasing hormone activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0