This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02598394_s1
See other H3F3C GE Assays ›
Gene Symbol:
H3F3C
Gene Name:
H3 histone, family 3C
Gene Aliases:
H3.5
Chromosome Location:
Chr.12: 31791185 - 31792241 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02598394_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H3F3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013699.2 NP_001013721.2 1 361 120
GenBank mRNA BC066906.1 - 1 361 5267388 120
HQ873957.1 - 1 361 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010299.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001368.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene H3F3C

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell growth
protein binding
nucleosomal DNA binding
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716282 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283610 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283562 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716280 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16546415 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716228 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283569 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716227 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283593 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716269 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716267 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16546397 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716248 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716222 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716245 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716285 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716251 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283564 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716240 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716226 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283614 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716230 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716223 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P16546408 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283575 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716284 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716274 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716275 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716235 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716217 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716218 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716239 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283621 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283594 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716236 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716231 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716242 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716232 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716229 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P28716215 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283600 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283608 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P08283618 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 122346 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P12214 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 30571 - -
Agilent Human Genome CGH + SNP Microarray A_18_P11398796 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P82419 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283614 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283608 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716267 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716266 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283605 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283597 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716271 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283600 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283621 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283610 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716279 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283618 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716269 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716251 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546407 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546405 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716265 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546398 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546410 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283600 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283608 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716282 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716269 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716276 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283621 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716267 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283618 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716284 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716248 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546415 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546410 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283605 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283614 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716277 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716270 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283597 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716265 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716271 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716268 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716275 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546398 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716266 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716251 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716279 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716285 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08283610 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546405 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28716280 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16546407 - -
Whole Human Genome Oligo Microarray G4112A A_32_P12214 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283621 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283608 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283612 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283610 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283605 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283599 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283601 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283616 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283602 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283604 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283597 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P08283614 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P08283600 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P08283601 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P12214 - -
Custom Human Agilent Array 11k, Chromosome 6, Expression 5433 - -
Agilent Human 1B Microarray G4111A A_32_P12214 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283565 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283568 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283583 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283618 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283575 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283622 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283589 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283608 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283588 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283610 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283621 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283571 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283596 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283612 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283599 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283592 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283563 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283579 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283572 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283594 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283614 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283567 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283570 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283616 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283586 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283601 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283604 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P08283606 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_001013699_at - -
Affymetrix Human Gene 1.0 ST Array 7962195 - -
Affymetrix Human Gene 1.0 ST Array 7962196 - -
Affymetrix Human Gene 1.1 ST Array 7962194 - -
Affymetrix Human Genome U219 Array 11715188_PM_at - -
Affymetrix Human Genome U219 Array 11715188_at - -
Affymetrix Human Transcriptome Array 2.0 PSR12017887.hg.1 ENST00000340398.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12017887.hg.1 NM_001013699.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12017887.hg.1 TC12001368.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12017885.hg.1 ENST00000340398.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12017885.hg.1 NM_001013699.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12017885.hg.1 TC12001368.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200178078.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178078.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178078.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178078.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178078.hg.1 TC1200010299.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200178079.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178079.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178079.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178079.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178079.hg.1 TC1200010299.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay TC1200010299.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay TC1200010299.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178077.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178077.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178077.hg.1 ENST00000340398.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200178077.hg.1 NM_001013699.2 Refseq
Affymetrix Human Clariom D Assay PSR1200178077.hg.1 TC1200010299.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A047458 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A047458 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 167176 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1769705 BC066906.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1769705 HQ873957.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2061979 BC066906.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_19743-S - -
Illumina human-6 v2.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip 4730100 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_19743 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769705 BC066906.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1769705 HQ873957.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2061979 HQ873957.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1769705 BC066906.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1769705 HQ873957.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2061979 BC066906.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2061979 HQ873957.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0