Assay Details
Gene Symbol: IGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152855.2 | NP_690594.1 | 1 - 2 | 324 | 111 | |
GenBank mRNA | BC030239.1 | - | 1 - 2 | 266 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000562.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phagocytosis, recognition
phagocytosis, engulfment
immune response
complement activation, classical pathway
defense response to bacterium
innate immune response
B cell receptor signaling pathway
positive regulation of B cell activation
antigen binding
immunoglobulin receptor binding
phagocytosis, recognition
phagocytosis, engulfment
immune response
complement activation, classical pathway
defense response to bacterium
innate immune response
B cell receptor signaling pathway
positive regulation of B cell activation
antigen binding
immunoglobulin receptor binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22010105.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010105.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010105.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010105.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010105.hg.1 | TC22000562.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | ENST00000412472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | ENST00000412472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010104.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010104.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010104.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010104.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010104.hg.1 | TC22000562.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008174.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158460.hg.1 | TC2200008174.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158462.hg.1 | TC2200008174.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | ENST00000438703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158463.hg.1 | TC2200008174.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | ENST00000249053.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | ENST00000330377.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | NM_020070.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | NM_152855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158461.hg.1 | TC2200008174.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | ENST00000412472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | ENST00000412472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006920.hg.1 | NR_029395.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 141999 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 171778 | - | - |
