Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000282486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000282488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000324196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000324210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000355460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000357472.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000461436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000463374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000485509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000485910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000492948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000498502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
ENST00000545754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_021038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207292.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207293.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207294.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
NM_207297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014850.hg.1 |
TC03000832.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000282486.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000282488.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000324196.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000324210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000355460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000357472.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000461436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000463374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000485509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000485910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000492948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000498502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
ENST00000545754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_021038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207292.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207293.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207294.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
NM_207297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014851.hg.1 |
TC03000832.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000282486.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000282488.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000324196.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000324210.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000355460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000357472.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000461436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000463374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000464596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000465907.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000485509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000485910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000492948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000498502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000545754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_021038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207292.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207293.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207294.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000282486.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000282488.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000324196.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000324210.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000355460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000357472.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000461436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000463374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000464596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000465907.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000485509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000485910.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000492948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000498502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
ENST00000545754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_021038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207292.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207293.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207294.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
NM_207297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300166044.hg.1 |
TC0300009202.hg.1 |
Clariom D TC |