This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02516756_s1
See other SLC28A1 GE Assays ›
Gene Symbol:
SLC28A1
Gene Name:
solute carrier family 28 member 1
Gene Aliases:
CNT1, HCNT1
Chromosome Location:
Chr.15: 84884351 - 84975649 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs02516756_s1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC28A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201651.2 NP_964014.1 7 1188 87
GenBank mRNA BC029788.1 - 7 1149 87
BC039898.1 - 6 1048 87
BI765705.1 - 1 448 87

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC28A1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
pyrimidine nucleobase transport
nucleoside transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
purine nucleobase transmembrane transport
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0