This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01906789_s1
See other KCND3 GE Assays ›
Gene Symbol:
KCND3
Gene Name:
potassium voltage-gated channel subfamily D member 3
Gene Aliases:
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22
Chromosome Location:
Chr.1: 111770662 - 111989577 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01906789_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCND3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005270851.4 XP_005270908.1 8 7467 122
XM_006710629.3 XP_006710692.1 8 7777 122
XM_006710630.3 XP_006710693.1 7 7410 122
XM_017001244.1 XP_016856733.1 8 7792 122
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA627904.1 - 1 Chr. 1 - 111770666 - 111771117 [+] on Build GRCh38 122
GenBankmRNA AA948623.1 - 1 Chr. 1 - 111770666 - 111770863 [+] on Build GRCh38 122
GenBankmRNA AI474561.1 - 1 Chr. 1 - 111770666 - 111770967 [+] on Build GRCh38 122
GenBankmRNA AI275874.1 - 1 Chr. 1 - 111770662 - 111771013 [+] on Build GRCh38 122
GenBankmRNA BM980893.1 - 1 Chr. 1 - 111770662 - 111771342 [+] on Build GRCh38 122
GenBankmRNA AA150776.1 - 1 Chr. 1 - 111770665 - 111770870 [+] on Build GRCh38 122
GenBankmRNA CB050759.1 - 1 Chr. 1 - 111770662 - 111770970 [+] on Build GRCh38 122
GenBankmRNA AF070632.1 - 2 1348 Chr. 1 - 111770662 - 111772569 [-] on Build GRCh38 122
GenBankmRNA DB316702.1 - 1 Chr. 1 - 111770666 - 111771229 [+] on Build GRCh38 122
GenBankmRNA AI559935.1 - 1 Chr. 1 - 111770673 - 111770937 [+] on Build GRCh38 122
GenBankmRNA R45514.1 - Chr. - - [-] on Build 122
GenBankmRNA DB317450.1 - 1 Chr. 1 - 111770673 - 111771190 [+] on Build GRCh38 123
GenBankmRNA BU726159.1 - 1 Chr. 1 - 111770662 - 111771204 [+] on Build GRCh38 122
GenBankmRNA AA136273.1 - 1 Chr. 1 - 111770666 - 111771025 [+] on Build GRCh38 122
GenBankmRNA CD105658.1 - 1 373 Chr. 1 - 111770666 - 111771090 [-] on Build GRCh38 122
GenBankmRNA AI088550.1 - 1 Chr. 1 - 111770665 - 111771104 [+] on Build GRCh38 122
GenBankmRNA AA564311.1 - 1 Chr. 1 - 111770666 - 111770960 [+] on Build GRCh38 122
GenBankmRNA AI279480.1 - 1 Chr. 1 - 111770662 - 111771035 [+] on Build GRCh38 122
GenBankmRNA AA530995.1 - 1 Chr. 1 - 111770671 - 111771220 [+] on Build GRCh38 122

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DEAD-box helicase 20 DDX20 DP103,
GEMIN3
-3.1kb Sense Chr. 1 - 111755568 - 111767577 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
regulation of ion transmembrane transport
protein homooligomerization
cardiac conduction
potassium ion export
cellular response to BMP stimulus
membrane repolarization
A-type (transient outward) potassium channel activity
delayed rectifier potassium channel activity
protein binding
ion channel binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000302127.4 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000315987.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 ENST00000369697.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_004980.4 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq
Affymetrix Human Clariom D Assay TC0100015288.hg.1 NM_172198.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0