This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01904465_s1
See other DKFZP434A062 GE Assays ›
Gene Symbol:
DKFZP434A062
Gene Name:
uncharacterized LOC26102
Gene Aliases:
-
Chromosome Location:
Chr.9: 136322302 - 136327323 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01904465_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DKFZP434A062
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026964.3 1 2626 122
GenBank mRNA AK127730.1 - 1 2626 122
AL080191.1 - 1 521 122

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 187 CCDC187 -15.4kb Sense Chr. 9 - 136267437 - 136306901 on NCBI Build GRCh38
Downstream G-protein signaling modulator 1 GPSM1 AGS3 153.0 Anti-sense Chr. 9 - 136327476 - 136359605 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001727.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007954 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358606 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358607 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358598 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007956 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007962 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007963 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007957 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007960 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P07007959 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358601 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358608 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P27358600 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 47551 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 62402 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 562443 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 547349 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 256232 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P667968 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4890 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P667968 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4891 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 41641 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 29519 - -
Agilent Human Genome CGH + SNP Microarray A_16_P18799710 - -
Agilent Human Genome CGH + SNP Microarray A_16_P18799710 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P667968 - -
SurePrint G3 Human GE Microarray G4851A A_24_P667968 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007959 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358608 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007960 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358629 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007954 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007968 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007971 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007962 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007966 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007978 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007972 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276568 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007963 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007970 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358623 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276569 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007974 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358625 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007967 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276570 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007957 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007980 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358606 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007978 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007959 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358629 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007970 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007974 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007960 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007972 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358625 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276569 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007957 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358606 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358623 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007963 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007967 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007968 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007966 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007962 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007980 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276570 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16276568 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007954 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27358608 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07007971 - -
Whole Human Genome Oligo Microarray G4112A A_24_P667968 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18799710 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18799710 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P667968 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 106360 - -
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 94053 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18799710 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32578013 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007973 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007966 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007955 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007971 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007978 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16276569 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007964 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007976 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007963 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007970 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16276565 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007957 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007980 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007954 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007974 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007960 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16276568 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07007972 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18799710 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 217219_PM_at - -
Affymetrix HT HG-U133+ PM Array Plate 216608_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09022063.hg.1 NR_026964.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09022063.hg.1 TC09001727.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900181941.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181941.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181941.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181941.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181941.hg.1 TC0900011973.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay TC0900011973.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay TC0900011973.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay TC0900011973.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay TC0900011973.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181940.hg.1 TC0900011973.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 ENST00000573103.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 ENST00000601276.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 NR_026964.3 Refseq
Affymetrix Human Clariom D Assay PSR0900181939.hg.1 TC0900011973.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 216608_at - -
Human Genome U133 Plus 2.0 Array 217219_PM_at - -
Human Genome U133 Plus 2.0 Array 217219_at - -
Human Genome U133 Plus 2.0 Array 216608_PM_at - -
Human Genome U133A 2.0 Array 217219_at - -
Human Genome U133A 2.0 Array 216608_PM_at - -
Human Genome U133A 2.0 Array 217219_PM_at - -
Human Genome U133A 2.0 Array 216608_at - -
AffymetrixPathwork Diagnostics Pathchip pw526741 - -
AffymetrixPathwork Diagnostics Pathchip pw599132 - -
Human Genome U95 Set 31660_PM_at - -
Human Genome U95 Set 31660_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 216608_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 217219_at7 - -
Human Genome U133 Set 216608_PM_at - -
Human Genome U133 Set 217219_at - -
Human Genome U133 Set 216608_at - -
Human Genome U133 Set 217219_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 234269 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_3266133 AL080191.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3266133 NR_026964.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_3266133 AK127730.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0