Assay Details
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020431.3 | NP_065164.2 | 19 - 20 | 1982 | 100 | |
GenBank mRNA | AK124749.1 | - | 5 - 6 | 490 | 100 | |
AK125159.1 | - | 19 - 20 | 1914 | 100 | ||
BC136613.1 | - | 19 - 20 | 1864 | 9020603 | 100 | |
BC136614.1 | - | 19 - 20 | 1861 | 9020604 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14002202.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14006201.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14006201.hg.1 | ENST00000536110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14006201.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14006201.hg.1 | TC14002202.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400180235.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400180235.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400180235.hg.1 | ENST00000298351.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400180235.hg.1 | NM_020431.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400180235.hg.1 | TC1400010635.hg.1 | Clariom D TC |
