This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01690600_g1
See other DPP10-AS1 GE Assays ›
Gene Symbol:
DPP10-AS1
Gene Name:
DPP10 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 115144048 - 115161343 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01690600_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPP10-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036580.1 2 - 3 407 72
GenBank mRNA AB593078.1 - 2 - 3 429 72
BC032913.2 - 2 - 3 407 72
BC048425.1 - 2 - 3 400 72
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG396693.1 - 2 - 3 417 Chr. 2 - 115144048 - 115161353 [-] on Build GRCh38 72
GenBankmRNA AI183963.1 - 1 - 2 Chr. 2 - 115144048 - 115144981 [+] on Build GRCh38 72
GenBankmRNA DB518722.1 - 1 - 2 144 Chr. 2 - 115144049 - 115145004 [-] on Build GRCh38 72
GenBankmRNA BI459080.1 - 2 - 3 412 Chr. 2 - 115144086 - 115161343 [-] on Build GRCh38 72
GenBankmRNA HY235889.1 - 1 - 2 Chr. 2 - 115144049 - 115145029 [+] on Build GRCh38 72
GenBankmRNA AW206823.1 - 1 - 2 Chr. 2 - 115144048 - 115144956 [+] on Build GRCh38 72
GenBankmRNA BE255017.1 - 2 - 3 396 Chr. 2 - 115144198 - 115161333 [-] on Build GRCh38 72
GenBankmRNA AA398846.1 - 2 - 3 253 Chr. 2 - 115144169 - 115161190 [-] on Build GRCh38 72
GenBankmRNA BG717164.1 - 2 - 3 410 Chr. 2 - 115144106 - 115161343 [-] on Build GRCh38 72
GenBankmRNA HY239423.1 - 1 - 2 Chr. 2 - 115144049 - 115144979 [+] on Build GRCh38 72
GenBankmRNA BG470876.1 - 2 - 3 416 Chr. 2 - 115144048 - 115161353 [-] on Build GRCh38 72
GenBankmRNA AA435891.1 - 1 - 2 Chr. 2 - 115144048 - 115144956 [+] on Build GRCh38 72
GenBankmRNA AI073569.1 - 1 - 2 Chr. 2 - 115144048 - 115161189 [+] on Build GRCh38 72
GenBankmRNA BX095397.1 - 2 - 3 253 Chr. 2 - 115144054 - 115161189 [-] on Build GRCh38 72
GenBankmRNA BE832976.1 - 1 - 2 Chr. 2 - 115144171 - 115144951 [+] on Build GRCh38 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DPP10 antisense RNA 3 DPP10-AS3 -310.5kb Sense Chr. 2 - 114828341 - 114833548 on NCBI Build GRCh38
Downstream DEAD-box helicase 18 DDX18 MrDb 2.7mb Anti-sense Chr. 2 - 117814679 - 117832377 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013974.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002240.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_32_P86578 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P86578 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 42005 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13672 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_32_P86578 - -
Agilent Human 1B Microarray G4111A A_32_P86578 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 236351_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036226.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036226.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036226.hg.1 TC02002240.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02036227.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036227.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02036227.hg.1 NR_036580.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02036227.hg.1 TC02002240.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay PSR0200202454.hg.1 TC0200013974.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 ENST00000448663.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay TC0200013974.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay PSR0200202453.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202453.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay PSR0200202453.hg.1 ENST00000432658.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200202453.hg.1 NR_036580.1 Refseq
Affymetrix Human Clariom D Assay PSR0200202453.hg.1 TC0200013974.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 236351_at - -
Human Genome U133 Plus 2.0 Array 236351_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 236351_at11 - -
Human Genome U95 Set 48987_PM_at - -
Human Genome U95 Set 48987_at - -
Human Genome U133 Set 236351_at - -
Human Genome U133 Set 236351_PM_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A014249 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A014249 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 192861 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0