This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01681301_m1
See other GBA2 GE Assays ›
Gene Symbol:
GBA2
Gene Name:
glucosylceramidase beta 2
Gene Aliases:
AD035, NLGase, SPG46
Chromosome Location:
Chr.9: 35736862 - 35749255 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01681301_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GBA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020944.2 NP_065995.1 1 - 2 883 128
XM_005251526.4 XP_005251583.1 1 - 2 910 128
XM_006716809.3 XP_006716872.1 1 - 2 910 128
XM_011517973.2 XP_011516275.1 1 - 2 488 128
XM_017014937.1 XP_016870426.1 1 - 2 910 128
XM_017014938.1 XP_016870427.1 1 - 2 910 128
XM_017014939.1 XP_016870428.1 1 - 2 488 128
XM_017014940.1 XP_016870429.1 1 - 2 488 128
XM_017014941.1 XP_016870430.1 1 - 2 910 128
XM_017014942.1 XP_016870431.1 1 - 2 488 128
XM_017014943.1 XP_016870432.1 1 - 2 488 128
XM_017014946.1 XP_016870435.1 1 - 2 488 128
GenBank mRNA AB046825.1 - 1 - 2 873 128
AJ309567.1 - 1 - 2 883 128
AK027884.1 - 1 - 2 546 128
AK295967.1 - 2 - 3 858 128
AL834306.1 - 1 - 2 643 128
BC011363.1 - 1 - 2 842 4339156 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009952.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001060.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017014942.1,XM_017014943.1,XM_017014946.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GBA2

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

glucosylceramide catabolic process
glycosphingolipid metabolic process
bile acid metabolic process
glycoside catabolic process
central nervous system neuron development
glucosylceramidase activity
beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09014187.hg.1 ENST00000378094.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09014187.hg.1 ENST00000378103.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09014187.hg.1 ENST00000545786.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09014187.hg.1 NM_020944.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09014187.hg.1 TC09001060.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000378094.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000378103.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000489025.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 NM_020944.2 Refseq
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000378094.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000378103.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 ENST00000489025.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 NM_020944.2 Refseq
Affymetrix Human Clariom D Assay PSR0900168559.hg.1 TC0900009952.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0