This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01651514_m1
See other NPSR1-AS1 GE Assays ›
Gene Symbol:
NPSR1-AS1
Gene Name:
NPSR1 antisense RNA 1
Gene Aliases:
AAA1
Chromosome Location:
Chr.7: 34346512 - 34834331 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01651514_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPSR1-AS1
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY312367.1 - LIT3327
AAA1
4 - 5 553 127
U85653.1 -
4 - 5 516 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000358772.4 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000419766.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000428922.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000431669.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000436945.3 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000439852.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000442669.2 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000535640.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000537560.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000539747.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 ENST00000544556.1 Ensembl
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_015356.2 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033664.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq
Affymetrix Human Clariom D Assay TC0700010710.hg.1 NR_033665.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0