Assay Details
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191013.1 | NP_001177942.1 | 1 - 2 | 394 | 177 | |
NM_183239.1 | NP_899062.1 | 1 - 2 | 394 | 177 | ||
GenBank mRNA | BC056918.1 | - | 1 - 2 | 394 | 5165084 | 177 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000782.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC056918.1,NM_001191013.1,NM_183239.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity
glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | ENST00000338595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | ENST00000369708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | ENST00000450629.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009335.hg.1 | TC10000782.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | ENST00000338595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | ENST00000369708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | ENST00000450629.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10009333.hg.1 | TC10000782.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186260.hg.1 | TC1000012498.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186261.hg.1 | TC1000012498.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186262.hg.1 | TC1000012498.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 177000 | - | - |
