This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01597868_m1
See other PHLPP1 GE Assays ›
Gene Symbol:
PHLPP1
Gene Name:
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases:
PHLPP, PLEKHE1, PPM3A, SCOP
Chromosome Location:
Chr.18: 62715439 - 62980443 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01597868_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHLPP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194449.3 NP_919431.2 16 - 17 4234 172
GenBank mRNA AB011178.2 - 16 - 17 2947 172
BC010706.1 - 3 - 4 469 172
BC014927.2 - 16 - 17 2521 3916044 172
BC047653.1 - 5 - 6 808 172
BC063519.1 - 4 - 5 682 172
BC126277.1 - 16 - 17 2569 8991993 172

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007523.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000224.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHLPP1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of T cell anergy
protein dephosphorylation
apoptotic process
entrainment of circadian clock
regulation of apoptotic process
regulation of MAPK cascade
regulation of JNK cascade
negative regulation of protein kinase B signaling
regulation of p38MAPK cascade
phosphoprotein phosphatase activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000262719.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000487409.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000497351.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000497351.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000497351.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000497351.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588452.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588452.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588452.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588452.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588452.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588953.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588953.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000588953.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591106.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 ENST00000591386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800007523.hg.1 NM_194449.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0