Assay Details
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168241.1 | NP_001161713.1 | 4 - 5 | 1553 | 119 | |
NM_001191033.1 | NP_001177962.1 | 3 - 4 | 2948 | 119 | ||
XM_006711951.3 | XP_006712014.1 | 4 - 5 | 1509 | 119 | ||
XM_011532564.2 | XP_011530866.1 | 4 - 5 | 1990 | 119 | ||
XM_011532565.2 | XP_011530867.1 | 5 - 6 | 1563 | 119 | ||
XM_011532566.2 | XP_011530868.1 | 4 - 5 | 1321 | 119 | ||
XM_011532567.2 | XP_011530869.1 | 4 - 5 | 1516 | 119 | ||
GenBank mRNA | AB124552.1 | - | 4 - 5 | 1553 | 119 | |
AK090454.1 | - | 3 - 4 | 2950 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000148.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02001692.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001692.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001692.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001692.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001692.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001687.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001687.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001687.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001688.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001688.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001688.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001686.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001686.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001686.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001690.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001690.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001690.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001690.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001690.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001691.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001689.hg.1 | ENST00000401533.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001689.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001689.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001689.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001689.hg.1 | TC02000148.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200147779.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147779.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147779.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147779.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147779.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147781.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147781.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147781.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147781.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147781.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147786.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147787.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | ENST00000496070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007028.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147782.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147782.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147782.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147782.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147782.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147784.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | ENST00000407684.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | NM_001191033.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147785.hg.1 | TC0200007028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200147780.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147780.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147780.hg.1 | ENST00000401533.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147780.hg.1 | NM_001168241.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147780.hg.1 | TC0200007028.hg.1 | Clariom D TC |
